Product Details

SNP ID
rs200312334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19878402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGATGCATCTCGTCCAGCCAC[C/T]GTGAACTCCAGTGGTTTATAATGGG
Phenotype
MIM: 606448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXNRD2 PubMed Links

Gene Details

Gene
TXNRD2
Gene Name
thioredoxin reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282512.1 1500 Intron NP_001269441.1
NM_006440.4 1500 Silent Mutation ACA,ACG T437T NP_006431.2

View Full Product Details