Product Details

SNP ID
rs200443600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30525050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGGTCTGGGAGCAGTACCTCCA[C/T]GGTGTAGCTGATGCGTTTAGAATGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEC14L6 PubMed Links

Gene Details

Gene
SEC14L6
Gene Name
SEC14 like lipid binding 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193336.2 1220 Missense Mutation NP_001180265.2
XM_011530357.2 1220 Missense Mutation XP_011528659.1
XM_011530358.2 1220 Missense Mutation XP_011528660.1
XM_011530360.2 1220 Missense Mutation XP_011528662.1
XM_017028931.1 1220 Missense Mutation XP_016884420.1
XM_017028932.1 1220 Missense Mutation XP_016884421.1
XM_017028933.1 1220 UTR 3 XP_016884422.1
XM_017028934.1 1220 UTR 3 XP_016884423.1

View Full Product Details