Product Details

SNP ID
rs201849924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38427014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCATAAGGGCCAGCTCGTTCTC[A/G]TAGCAGAAGGCACTGGGAGGGGGCG
Phenotype
MIM: 600504
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNJ4 PubMed Links

Gene Details

Gene
KCNJ4
Gene Name
potassium voltage-gated channel subfamily J member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004981.1 1216 Silent Mutation TAC,TAT Y373Y NP_004972.1
NM_152868.2 1216 Silent Mutation TAC,TAT Y373Y NP_690607.1
Gene
LOC101927183
Gene Name
uncharacterized LOC101927183
There are no transcripts associated with this gene.

View Full Product Details