Product Details

SNP ID
rs200522444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31105259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCACGAGCTCAGGGTCGGCCCC[A/G]GGGAGGTGTTTCATCACCAGGTTGT
Phenotype
MIM: 606481 MIM: 610918 MIM: 602127
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INPP5J PubMed Links

Gene Details

Gene
INPP5J
Gene Name
inositol polyphosphate-5-phosphatase J
There are no transcripts associated with this gene.

Gene
SELM
Gene Name
selenoprotein M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080430.2 288 Silent Mutation CCC,CCT P76P NP_536355.1
Gene
SMTN
Gene Name
smoothelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207017.1 288 Intron NP_001193946.1
NM_001207018.1 288 Intron NP_001193947.1
NM_006932.4 288 Intron NP_008863.3
NM_134269.2 288 Intron NP_599031.1
NM_134270.2 288 Intron NP_599032.2
XM_006724298.1 288 Intron XP_006724361.1
XM_006724302.1 288 Intron XP_006724365.1
XM_011530333.2 288 Intron XP_011528635.1
XM_011530336.2 288 Intron XP_011528638.1
XM_011530337.2 288 Intron XP_011528639.1
XM_011530338.2 288 Intron XP_011528640.1
XM_011530339.2 288 Intron XP_011528641.1
XM_011530340.2 288 Intron XP_011528642.1
XM_011530341.1 288 Intron XP_011528643.1
XM_017028913.1 288 Intron XP_016884402.1
XM_017028914.1 288 Intron XP_016884403.1
XM_017028915.1 288 Intron XP_016884404.1
XM_017028916.1 288 Intron XP_016884405.1
XM_017028917.1 288 Intron XP_016884406.1
XM_017028918.1 288 Intron XP_016884407.1
XM_017028919.1 288 Intron XP_016884408.1

View Full Product Details