Product Details

SNP ID
rs201243203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24586226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGGCGGGGCAGCGCCCACAGG[A/C]TGGGCAGCAGGAGGTGCAGCAGCTC
Phenotype
MIM: 612346
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GGT1 PubMed Links

Gene Details

Gene
GGT1
Gene Name
gamma-glutamyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288833.1 637 Intron NP_001275762.1
NM_013421.2 637 Intron NP_038265.2
NM_013430.2 637 Intron NP_038347.2
Gene
LRRC75B
Gene Name
leucine rich repeat containing 75B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207644.2 637 Missense Mutation AGC,ATC S203I NP_997527.2
XM_005261600.2 637 Intron XP_005261657.1

View Full Product Details