Product Details

SNP ID
rs200140395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50277615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGCTCAGCTTATGCTCCGTGCG[C/T]GTGCAGGCATCCATGAAGGTCTGCG
Phenotype
MIM: 602898 MIM: 604293
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPK11 PubMed Links

Gene Details

Gene
MAPK11
Gene Name
mitogen-activated protein kinase 11
There are no transcripts associated with this gene.

Gene
PLXNB2
Gene Name
plexin B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012401.3 5265 Silent Mutation ACA,ACG T1724T NP_036533.2
XM_005261909.1 5265 Silent Mutation ACA,ACG T1724T XP_005261966.1
XM_005261910.2 5265 Silent Mutation ACA,ACG T1724T XP_005261967.1
XM_005261911.4 5265 Silent Mutation ACA,ACG T1724T XP_005261968.1
XM_006724413.2 5265 Silent Mutation ACA,ACG T1724T XP_006724476.1
XM_011530682.2 5265 Silent Mutation ACA,ACG T1787T XP_011528984.1
XM_017028703.1 5265 Silent Mutation ACA,ACG T1724T XP_016884192.1
XM_017028704.1 5265 Silent Mutation ACA,ACG T1724T XP_016884193.1

View Full Product Details