Product Details
- SNP ID
-
rs200814486
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:138670943 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGATCAGCCAGAAGATAAGTATGG[A/G]GTCTCACTATGGTAATTGTAGTCAG
- Phenotype
-
MIM: 604635
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NXPH2
PubMed Links
Gene Details
- Gene
- NXPH2
- Gene Name
- neurexophilin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007226.2 |
774 |
Silent Mutation |
ACC,ACT |
T258T |
NP_009157.1 |
View Full Product Details