Product Details

SNP ID
rs200789364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112832728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCATATGGACCAGACATCACCA[A/G]GCTTTTTTGCTGTGAGTCCCGGAGC
Phenotype
MIM: 147720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL1B PubMed Links

Gene Details

Gene
IL1B
Gene Name
interleukin 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000576.2 487 Silent Mutation CTG,TTG L134L NP_000567.1
XM_017003988.1 487 Silent Mutation CTG,TTG L103L XP_016859477.1

View Full Product Details