Product Details

SNP ID
rs200655333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197489224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCTGAACGTCTTCAAGATTCAG[C/G]GTCAATCCCTCTTCTCCAAACACCT
Phenotype
MIM: 118190
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HSPD1 PubMed Links

Gene Details

Gene
HSPD1
Gene Name
heat shock protein family D (Hsp60) member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002156.4 Intron NP_002147.2
NM_199440.1 Intron NP_955472.1
Gene
SNORA105B
Gene Name
small nucleolar RNA, H/ACA box 105B
There are no transcripts associated with this gene.

View Full Product Details