Product Details

SNP ID
rs201258584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73230150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTCCTTGGCAGGTGTGGCTGTG[A/G]AGATGTATCGAATGTCCCCAGGACT
Phenotype
MIM: 605140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCT7 PubMed Links

Gene Details

Gene
CCT7
Gene Name
chaperonin containing TCP1 subunit 7
There are no transcripts associated with this gene.

Gene
PRADC1
Gene Name
protease associated domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032319.1 199 Missense Mutation TCC,TTC S44F NP_115695.1
Gene
SMYD5
Gene Name
SMYD family member 5
There are no transcripts associated with this gene.

View Full Product Details