Product Details
- SNP ID
-
rs200781797
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241495666 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATCTGAACAGCAGTGCGCTTCAG[C/T]GGGTGGATGTCAGGTGGTTTCTGTT
- Phenotype
-
MIM: 602255
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FARP2
PubMed Links
Gene Details
- Gene
- FARP2
- Gene Name
- FERM, ARH/RhoGEF and pleckstrin domain protein 2
- Gene
- STK25
- Gene Name
- serine/threonine kinase 25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001271977.1 |
1336 |
Missense Mutation |
CAC,CGC |
H426R |
NP_001258906.1 |
NM_001271978.1 |
1336 |
Missense Mutation |
CAC,CGC |
H426R |
NP_001258907.1 |
NM_001271979.1 |
1336 |
Missense Mutation |
CAC,CGC |
H349R |
NP_001258908.1 |
NM_001271980.1 |
1336 |
Missense Mutation |
CAC,CGC |
H349R |
NP_001258909.1 |
NM_001282305.1 |
1336 |
Missense Mutation |
CAC,CGC |
H332R |
NP_001269234.1 |
NM_001282306.1 |
1336 |
Missense Mutation |
CAC,CGC |
H352R |
NP_001269235.1 |
NM_001282307.1 |
1336 |
Missense Mutation |
CAC,CGC |
H332R |
NP_001269236.1 |
NM_001282308.1 |
1336 |
Missense Mutation |
CAC,CGC |
H332R |
NP_001269237.1 |
NM_006374.4 |
1336 |
Missense Mutation |
CAC,CGC |
H426R |
NP_006365.2 |
XM_011510493.2 |
1336 |
Intron |
|
|
XP_011508795.1 |
XM_011510494.2 |
1336 |
Missense Mutation |
CAC,CGC |
H352R |
XP_011508796.1 |
XM_011510495.2 |
1336 |
Missense Mutation |
CAC,CGC |
H352R |
XP_011508797.1 |
XM_011510496.2 |
1336 |
Intron |
|
|
XP_011508798.1 |
XM_017003168.1 |
1336 |
UTR 3 |
|
|
XP_016858657.1 |
XM_017003169.1 |
1336 |
UTR 3 |
|
|
XP_016858658.1 |
XM_017003170.1 |
1336 |
Missense Mutation |
CAC,CGC |
H352R |
XP_016858659.1 |
XM_017003171.1 |
1336 |
UTR 3 |
|
|
XP_016858660.1 |
View Full Product Details