Product Details

SNP ID
rs201221821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:66437746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTTTGTCATGCAGTACGACGAT[C/T]TACCCCATTACGGGGGCATGGATGG
Phenotype
MIM: 601739
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEIS1 PubMed Links

Gene Details

Gene
MEIS1
Gene Name
Meis homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002398.2 802 Silent Mutation CTA,TTA L8L NP_002389.1
XM_005264321.2 802 Silent Mutation CTA,TTA L24L XP_005264378.1
XM_005264322.2 802 Silent Mutation CTA,TTA L8L XP_005264379.1
XM_005264323.2 802 Silent Mutation CTA,TTA L24L XP_005264380.1
XM_005264324.4 802 Intron XP_005264381.1
XM_005264325.4 802 Intron XP_005264382.1
XM_017004141.1 802 Silent Mutation CTA,TTA L24L XP_016859630.1
XM_017004142.1 802 Silent Mutation CTA,TTA L8L XP_016859631.1
XM_017004143.1 802 Intron XP_016859632.1
XM_017004144.1 802 UTR 5 XP_016859633.1
XM_017004145.1 802 UTR 5 XP_016859634.1
XM_017004146.1 802 UTR 5 XP_016859635.1
Gene
MEIS1-AS2
Gene Name
MEIS1 antisense RNA 2
There are no transcripts associated with this gene.

Gene
MEIS1-AS3
Gene Name
MEIS1 antisense RNA 3
There are no transcripts associated with this gene.

View Full Product Details