Product Details
- SNP ID
-
rs200894546
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:218638604 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGCCAGCAGCCCAGCGGAAAGCA[C/T]GGTGGCACACATTGCACACATAGGG
- Phenotype
-
MIM: 605939
MIM: 604083
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLCD4
PubMed Links
Gene Details
- Gene
- PLCD4
- Gene Name
- phospholipase C delta 4
There are no transcripts associated with this gene.
- Gene
- ZNF142
- Gene Name
- zinc finger protein 142
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001105537.2 |
6007 |
Missense Mutation |
CAT,CGT |
H1600R |
NP_001099007.1 |
XM_006712730.3 |
6007 |
Missense Mutation |
CAT,CGT |
H1800R |
XP_006712793.1 |
XM_011511784.2 |
6007 |
Missense Mutation |
CAT,CGT |
H1800R |
XP_011510086.1 |
XM_011511785.2 |
6007 |
Missense Mutation |
CAT,CGT |
H1800R |
XP_011510087.1 |
XM_011511786.2 |
6007 |
Missense Mutation |
CAT,CGT |
H1800R |
XP_011510088.1 |
XM_011511787.2 |
6007 |
Missense Mutation |
CAT,CGT |
H1800R |
XP_011510089.1 |
XM_011511788.2 |
6007 |
Missense Mutation |
CAT,CGT |
H1800R |
XP_011510090.1 |
XM_011511789.2 |
6007 |
Missense Mutation |
CAT,CGT |
H1600R |
XP_011510091.1 |
XM_017004871.1 |
6007 |
Missense Mutation |
CAT,CGT |
H1800R |
XP_016860360.1 |
XM_017004872.1 |
6007 |
Missense Mutation |
CAT,CGT |
H1600R |
XP_016860361.1 |
View Full Product Details