Product Details

SNP ID
rs201035087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166198972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAATGACAGTAGCAGACACATC[C/G]TCTTGTTTCCGTTTTAGTGTGGTTG
Phenotype
MIM: 603415
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 5942 Missense Mutation GAC,GAG D1878E NP_002968.1
XM_005246757.2 5942 Missense Mutation GAC,GAG D1889E XP_005246814.1
XM_011511616.2 5942 Missense Mutation GAC,GAG D1889E XP_011509918.1
XM_011511617.2 5942 Missense Mutation GAC,GAG D1889E XP_011509919.1
XM_011511618.2 5942 Missense Mutation GAC,GAG D1878E XP_011509920.1
XM_011511619.2 5942 Intron XP_011509921.1
XM_017004668.1 5942 Missense Mutation GAC,GAG D1760E XP_016860157.1
XM_017004669.1 5942 Missense Mutation GAC,GAG D1641E XP_016860158.1

View Full Product Details