Product Details

SNP ID
rs199925077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219549780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTGAGCTCCTTCATGGCCTCA[A/C]CCACAGATGCGGACTATGAGTATGG
Phenotype
MIM: 610405 MIM: 610991
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHPF PubMed Links

Gene Details

Gene
CHPF
Gene Name
chondroitin polymerizing factor
There are no transcripts associated with this gene.

Gene
MIR3132
Gene Name
microRNA 3132
There are no transcripts associated with this gene.

Gene
OBSL1
Gene Name
obscurin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173408.1 1420 Intron NP_001166879.1
NM_001173431.1 1420 Intron NP_001166902.1
NM_015311.2 1420 Intron NP_056126.1
XM_005246424.4 1420 Intron XP_005246481.1
XM_005246427.4 1420 Intron XP_005246484.1
XM_011510857.2 1420 Intron XP_011509159.1
XM_011510863.2 1420 Intron XP_011509165.1
XM_011510864.2 1420 Intron XP_011509166.1
XM_011510865.2 1420 Intron XP_011509167.1
XM_011510866.2 1420 Intron XP_011509168.1
XM_017003696.1 1420 Intron XP_016859185.1
XM_017003697.1 1420 Intron XP_016859186.1
XM_017003698.1 1420 Intron XP_016859187.1
XM_017003699.1 1420 Intron XP_016859188.1
XM_017003700.1 1420 Intron XP_016859189.1
Gene
TMEM198
Gene Name
transmembrane protein 198
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005209.2 1420 Missense Mutation ACC,CCC T337P NP_001005209.1
NM_001303098.1 1420 Missense Mutation ACC,CCC T337P NP_001290027.1
XM_017003367.1 1420 Missense Mutation ACC,CCC T337P XP_016858856.1

View Full Product Details