Product Details

SNP ID
rs199817813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130341000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGAGGGGCCTCAGAGGACTCC[A/G]GTTCTGGGGTCTTAGTGGGGCCCTT
Phenotype
MIM: 613734 MIM: 612981
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC115 PubMed Links

Gene Details

Gene
CCDC115
Gene Name
coiled-coil domain containing 115
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321118.1 297 Missense Mutation CCG,CTG P108L NP_001308047.1
NM_001321119.1 297 Missense Mutation CCG,CTG P65L NP_001308048.1
NM_032357.3 297 Missense Mutation CCG,CTG P113L NP_115733.2
Gene
IMP4
Gene Name
IMP4 homolog, U3 small nucleolar ribonucleoprotein
There are no transcripts associated with this gene.

View Full Product Details