Product Details

SNP ID
rs199805770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216414776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGACAAAAGGCTCTGGCCCGCAG[A/G]GCTGAGAAGTTATTGGCAGAACAGC
Phenotype
MIM: 606622
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMARCAL1 PubMed Links

Gene Details

Gene
SMARCAL1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127207.1 216 Silent Mutation AGA,AGG R24R NP_001120679.1
NM_014140.3 216 Silent Mutation AGA,AGG R24R NP_054859.2
XM_005246631.2 216 Silent Mutation AGA,AGG R24R XP_005246688.1
XM_005246632.1 216 Silent Mutation AGA,AGG R24R XP_005246689.1
XM_017004228.1 216 UTR 5 XP_016859717.1

View Full Product Details