Product Details
- SNP ID
-
rs200087086
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:25383866 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTCCCATTCTGCATCTTCTCTT[C/T]TTCTTCCTCTGCACCTTCCTCTGCT
- Phenotype
-
MIM: 602415
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DTNB
PubMed Links
Gene Details
- Gene
- DTNB
- Gene Name
- dystrobrevin beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256303.1 |
1932 |
Missense Mutation |
AAA,GAA |
K610E |
NP_001243232.1 |
NM_001256304.1 |
1932 |
Intron |
|
|
NP_001243233.1 |
NM_001256308.2 |
1932 |
Intron |
|
|
NP_001243237.1 |
NM_001320932.1 |
1932 |
Intron |
|
|
NP_001307861.1 |
NM_001320933.1 |
1932 |
Missense Mutation |
AAA,GAA |
K580E |
NP_001307862.1 |
NM_001320934.1 |
1932 |
Missense Mutation |
AAA,GAA |
K587E |
NP_001307863.1 |
NM_001320935.1 |
1932 |
Missense Mutation |
AAA,GAA |
K406E |
NP_001307864.1 |
NM_001320936.1 |
1932 |
Intron |
|
|
NP_001307865.1 |
NM_001320937.1 |
1932 |
Intron |
|
|
NP_001307866.1 |
NM_021907.4 |
1932 |
Missense Mutation |
AAA,GAA |
K617E |
NP_068707.1 |
NM_033147.3 |
1932 |
Missense Mutation |
AAA,GAA |
K587E |
NP_149159.2 |
NM_033148.3 |
1932 |
Missense Mutation |
AAA,GAA |
K557E |
NP_149160.1 |
NM_183360.2 |
1932 |
Intron |
|
|
NP_899204.1 |
NM_183361.2 |
1932 |
Missense Mutation |
AAA,GAA |
K550E |
NP_899205.1 |
XM_005264182.1 |
1932 |
Intron |
|
|
XP_005264239.1 |
XM_011532671.1 |
1932 |
Missense Mutation |
AAA,GAA |
K623E |
XP_011530973.1 |
XM_011532672.2 |
1932 |
Intron |
|
|
XP_011530974.1 |
XM_011532673.1 |
1932 |
Missense Mutation |
AAA,GAA |
K616E |
XP_011530975.1 |
XM_011532674.1 |
1932 |
Intron |
|
|
XP_011530976.1 |
XM_011532675.1 |
1932 |
Intron |
|
|
XP_011530977.1 |
XM_011532676.1 |
1932 |
Missense Mutation |
AAA,GAA |
K593E |
XP_011530978.1 |
XM_011532677.2 |
1932 |
Missense Mutation |
AAA,GAA |
K566E |
XP_011530979.1 |
XM_011532679.1 |
1932 |
Intron |
|
|
XP_011530981.1 |
XM_011532680.2 |
1932 |
Missense Mutation |
AAA,GAA |
K476E |
XP_011530982.1 |
XM_011532681.2 |
1932 |
Missense Mutation |
AAA,GAA |
K470E |
XP_011530983.1 |
XM_017003529.1 |
1932 |
Missense Mutation |
AAA,GAA |
K586E |
XP_016859018.1 |
XM_017003530.1 |
1932 |
Intron |
|
|
XP_016859019.1 |
XM_017003531.1 |
1932 |
Intron |
|
|
XP_016859020.1 |
XM_017003532.1 |
1932 |
Intron |
|
|
XP_016859021.1 |
XM_017003533.1 |
1932 |
Intron |
|
|
XP_016859022.1 |
XM_017003534.1 |
1932 |
Intron |
|
|
XP_016859023.1 |
XM_017003535.1 |
1932 |
Intron |
|
|
XP_016859024.1 |
XM_017003536.1 |
1932 |
Intron |
|
|
XP_016859025.1 |
XM_017003537.1 |
1932 |
Missense Mutation |
AAA,GAA |
K485E |
XP_016859026.1 |
XM_017003538.1 |
1932 |
Missense Mutation |
AAA,GAA |
K463E |
XP_016859027.1 |
XM_017003539.1 |
1932 |
Missense Mutation |
AAA,GAA |
K446E |
XP_016859028.1 |
XM_017003540.1 |
1932 |
Missense Mutation |
AAA,GAA |
K440E |
XP_016859029.1 |
XM_017003541.1 |
1932 |
Intron |
|
|
XP_016859030.1 |
XM_017003542.1 |
1932 |
Missense Mutation |
AAA,GAA |
K389E |
XP_016859031.1 |
XM_017003543.1 |
1932 |
Missense Mutation |
AAA,GAA |
K389E |
XP_016859032.1 |
XM_017003544.1 |
1932 |
Missense Mutation |
AAA,GAA |
K383E |
XP_016859033.1 |
XM_017003545.1 |
1932 |
Missense Mutation |
AAA,GAA |
K383E |
XP_016859034.1 |
View Full Product Details