Product Details

SNP ID
rs200100676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95275937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTGCTGCCTGCCCAGGTGGTGC[G/T]GTACGAGGCGGGCTACGTGGTATGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PROM2 PubMed Links

Gene Details

Gene
PROM2
Gene Name
prominin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165977.2 424 Missense Mutation CGG,CTG R101L NP_001159449.1
NM_001165978.2 424 Missense Mutation CGG,CTG R101L NP_001159450.1
NM_001321070.1 424 UTR 5 NP_001307999.1
NM_144707.3 424 Missense Mutation CGG,CTG R101L NP_653308.2
XM_006712301.2 424 Intron XP_006712364.1
XM_011510672.2 424 Missense Mutation CGG,CTG R101L XP_011508974.1

View Full Product Details