Product Details
- SNP ID
-
rs199700349
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:70212723 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAGATTCTGGAGTCCTTATTCAC[G/T]GGGTTTCATACCCTGCCACTCGATA
- Phenotype
-
MIM: 603518
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TIA1
PubMed Links
Gene Details
- Gene
- TIA1
- Gene Name
- TIA1 cytotoxic granule-associated RNA binding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022037.2 |
1381 |
Missense Mutation |
CAG,CCG |
Q375P |
NP_071320.2 |
NM_022173.2 |
1381 |
Missense Mutation |
CAG,CCG |
Q386P |
NP_071505.2 |
XM_005264527.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q374P |
XP_005264584.1 |
XM_005264528.4 |
1381 |
Missense Mutation |
CAG,CCG |
Q321P |
XP_005264585.1 |
XM_005264531.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q285P |
XP_005264588.1 |
XM_011533077.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q385P |
XP_011531379.1 |
XM_011533078.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q286P |
XP_011531380.1 |
XM_011533081.2 |
1381 |
Missense Mutation |
CAG,CCG |
Q286P |
XP_011531383.1 |
XM_011533082.2 |
1381 |
Missense Mutation |
CAG,CCG |
Q286P |
XP_011531384.1 |
XM_017004789.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q349P |
XP_016860278.1 |
XM_017004790.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q338P |
XP_016860279.1 |
XM_017004791.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q310P |
XP_016860280.1 |
XM_017004792.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q286P |
XP_016860281.1 |
XM_017004793.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q286P |
XP_016860282.1 |
XM_017004794.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q286P |
XP_016860283.1 |
XM_017004795.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q285P |
XP_016860284.1 |
XM_017004796.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q246P |
XP_016860285.1 |
XM_017004797.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q246P |
XP_016860286.1 |
XM_017004798.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q246P |
XP_016860287.1 |
XM_017004799.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q246P |
XP_016860288.1 |
XM_017004800.1 |
1381 |
Missense Mutation |
CAG,CCG |
Q246P |
XP_016860289.1 |
View Full Product Details