Product Details
- SNP ID
-
rs200567322
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:96711693 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGTGATGGAGGAGGTGCCGAAGTA[C/G]TAGCCGCGGGGCAGGCGGACTCCGG
- Phenotype
-
MIM: 609552
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FER1L5
PubMed Links
Gene Details
- Gene
- FER1L5
- Gene Name
- fer-1 like family member 5
There are no transcripts associated with this gene.
- Gene
- LMAN2L
- Gene Name
- lectin, mannose binding 2 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001142292.1 |
680 |
Nonsense Mutation |
TAC,TAG |
Y260* |
NP_001135764.1 |
NM_001322346.1 |
680 |
Nonsense Mutation |
TAC,TAG |
Y115* |
NP_001309275.1 |
NM_001322347.1 |
680 |
Nonsense Mutation |
TAC,TAG |
Y122* |
NP_001309276.1 |
NM_001322350.1 |
680 |
Nonsense Mutation |
TAC,TAG |
Y111* |
NP_001309279.1 |
NM_001322351.1 |
680 |
Nonsense Mutation |
TAC,TAG |
Y104* |
NP_001309280.1 |
NM_001322352.1 |
680 |
Nonsense Mutation |
TAC,TAG |
Y122* |
NP_001309281.1 |
NM_001322354.1 |
680 |
Nonsense Mutation |
TAC,TAG |
Y115* |
NP_001309283.1 |
NM_001322355.1 |
680 |
Nonsense Mutation |
TAC,TAG |
Y104* |
NP_001309284.1 |
NM_001322356.1 |
680 |
Nonsense Mutation |
TAC,TAG |
Y104* |
NP_001309285.1 |
NM_030805.3 |
680 |
Nonsense Mutation |
TAC,TAG |
Y249* |
NP_110432.1 |
View Full Product Details