Product Details
- SNP ID
-
rs200231230
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:99018616 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGTGTGACAACATTTCTGAACTG[A/G]GCCACATCTCTACTCTGGATGGCCC
- Phenotype
-
MIM: 607166
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSGA10
PubMed Links
Gene Details
- Gene
- TSGA10
- Gene Name
- testis specific 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025244.2 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
NP_079520.1 |
NM_182911.3 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
NP_878915.2 |
XM_005264023.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_005264080.1 |
XM_005264024.2 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_005264081.1 |
XM_005264025.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_005264082.1 |
XM_005264026.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_005264083.1 |
XM_005264027.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_005264084.1 |
XM_005264029.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_005264086.1 |
XM_006712781.2 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_006712844.1 |
XM_011511934.1 |
2224 |
Silent Mutation |
GCC,GCT |
A814A |
XP_011510236.1 |
XM_011511935.2 |
2224 |
Silent Mutation |
GCC,GCT |
A742A |
XP_011510237.1 |
XM_011511937.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_011510239.1 |
XM_011511939.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_011510241.1 |
XM_011511940.2 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_011510242.1 |
XM_011511941.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_011510243.1 |
XM_011511942.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_011510244.1 |
XM_011511943.2 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_011510245.1 |
XM_017005035.1 |
2224 |
Silent Mutation |
GCC,GCT |
A809A |
XP_016860524.1 |
XM_017005036.1 |
2224 |
Intron |
|
|
XP_016860525.1 |
XM_017005037.1 |
2224 |
UTR 3 |
|
|
XP_016860526.1 |
XM_017005038.1 |
2224 |
Intron |
|
|
XP_016860527.1 |
XM_017005039.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_016860528.1 |
XM_017005040.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_016860529.1 |
XM_017005041.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_016860530.1 |
XM_017005042.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_016860531.1 |
XM_017005043.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_016860532.1 |
XM_017005044.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_016860533.1 |
XM_017005045.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_016860534.1 |
XM_017005046.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_016860535.1 |
XM_017005047.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_016860536.1 |
XM_017005048.1 |
2224 |
Silent Mutation |
GCC,GCT |
A614A |
XP_016860537.1 |
View Full Product Details