Product Details

SNP ID
rs200429101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119681354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACCTGGAAAGCAGTACCACTGC[C/T]GTGCACGCCCTGCTGGCCCCAGCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP221 PubMed Links

Gene Details

Gene
CFAP221
Gene Name
cilia and flagella associated protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271049.1 633 Intron NP_001257978.1
XM_006712353.3 633 Intron XP_006712416.1
XM_017003550.1 633 Intron XP_016859039.1
XM_017003551.1 633 Intron XP_016859040.1
XM_017003552.1 633 Intron XP_016859041.1
XM_017003553.1 633 Intron XP_016859042.1
XM_017003554.1 633 Intron XP_016859043.1
XM_017003555.1 633 Intron XP_016859044.1
XM_017003556.1 633 Intron XP_016859045.1
XM_017003557.1 633 Intron XP_016859046.1
XM_017003558.1 633 Intron XP_016859047.1
XM_017003559.1 633 Intron XP_016859048.1
Gene
TMEM177
Gene Name
transmembrane protein 177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105198.1 633 Silent Mutation GCC,GCT A167A NP_001098668.1
NM_001105199.1 633 Silent Mutation GCC,GCT A167A NP_001098669.1
NM_030577.2 633 Silent Mutation GCC,GCT A167A NP_085054.2

View Full Product Details