Product Details
- SNP ID
-
rs201699531
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:240888379 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGGAGCGCCTTGAGCCCGGAGCC[C/G]ATGCGCGGCGGGGGGCTGCGGCCCA
- Phenotype
-
MIM: 604285
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AGXT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11899555] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGXT
- Gene Name
- alanine-glyoxylate aminotransferase
There are no transcripts associated with this gene.
- Gene
- C2orf54
- Gene Name
- chromosome 2 open reading frame 54
View Full Product Details