Product Details

SNP ID
rs202007589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206176722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTATTGAACTCCACAGTGTCCC[A/G]GAAGTACAGGGCAGGACCGCCAATT
Phenotype
MIM: 600239
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR1 PubMed Links

Gene Details

Gene
GPR1
Gene Name
G protein-coupled receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098199.1 750 Missense Mutation CGG,TGG R176W NP_001091669.1
NM_001261452.1 750 Missense Mutation CGG,TGG R176W NP_001248381.1
NM_001261453.1 750 Missense Mutation CGG,TGG R176W NP_001248382.1
NM_001261454.1 750 Missense Mutation CGG,TGG R176W NP_001248383.1
NM_001261455.1 750 Missense Mutation CGG,TGG R176W NP_001248384.1
NM_005279.3 750 Missense Mutation CGG,TGG R176W NP_005270.2
XM_005246471.2 750 Missense Mutation CGG,TGG R176W XP_005246528.1
XM_017003832.1 750 Missense Mutation CGG,TGG R176W XP_016859321.1

View Full Product Details