Product Details

SNP ID
rs201465267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:8730862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCATGCTTTTTAGATTCTCATTC[A/T]GAATAGTGGTTGGGTTGGGACTGGA
Phenotype
MIM: 615759
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIDINS220 PubMed Links

Gene Details

Gene
KIDINS220
Gene Name
kinase D-interacting substrate 220kDa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020738.2 5356 Missense Mutation CAG,CTG Q1725L NP_065789.1

View Full Product Details