Product Details
- SNP ID
-
rs201896678
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:200871456 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTAATGTGTACATCATTAAGAGGT[C/T]GGTATGTCTTCTCATTTACTGGCAA
- Phenotype
-
MIM: 601951
MIM: 615811
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLK1
PubMed Links
Gene Details
- Gene
- CLK1
- Gene Name
- CDC like kinase 1
There are no transcripts associated with this gene.
- Gene
- PPIL3
- Gene Name
- peptidylprolyl isomerase like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032472.3 |
688 |
Missense Mutation |
CAA,CGA |
Q146R |
NP_115861.1 |
NM_130906.2 |
688 |
Missense Mutation |
CAA,CGA |
Q142R |
NP_570981.1 |
XM_005246651.3 |
688 |
Missense Mutation |
CAA,CGA |
Q146R |
XP_005246708.1 |
XM_005246652.4 |
688 |
Missense Mutation |
CAA,CGA |
Q142R |
XP_005246709.1 |
XM_011511358.2 |
688 |
Missense Mutation |
CAA,CGA |
Q142R |
XP_011509660.1 |
XM_017004352.1 |
688 |
Missense Mutation |
CAA,CGA |
Q146R |
XP_016859841.1 |
XM_017004353.1 |
688 |
Missense Mutation |
CAA,CGA |
Q146R |
XP_016859842.1 |
XM_017004354.1 |
688 |
Intron |
|
|
XP_016859843.1 |
XM_017004355.1 |
688 |
Intron |
|
|
XP_016859844.1 |
XM_017004356.1 |
688 |
Intron |
|
|
XP_016859845.1 |
XM_017004357.1 |
688 |
Missense Mutation |
CAA,CGA |
Q93R |
XP_016859846.1 |
XM_017004358.1 |
688 |
Missense Mutation |
CAA,CGA |
Q93R |
XP_016859847.1 |
XM_017004359.1 |
688 |
Missense Mutation |
CAA,CGA |
Q93R |
XP_016859848.1 |
XM_017004360.1 |
688 |
Missense Mutation |
CAA,CGA |
Q93R |
XP_016859849.1 |
View Full Product Details