Product Details

SNP ID
rs201498828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27212400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCACGACCCGGGTAGTCTTACGA[C/T]CCTGGTGCCCTGGGCTGCCGCCCTG
Phenotype
MIM: 114010 MIM: 604024
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATRAID PubMed Links

Gene Details

Gene
ATRAID
Gene Name
all-trans retinoic acid induced differentiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170795.1 370 Missense Mutation ACC,ATC T11I NP_001164266.1
NM_016085.4 370 UTR 5 NP_057169.2
NM_080592.3 370 Missense Mutation ACC,ATC T66I NP_542159.3
Gene
CAD
Gene Name
carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
There are no transcripts associated with this gene.

Gene
SLC5A6
Gene Name
solute carrier family 5 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021095.2 370 Intron NP_066918.2
XM_006712128.1 370 Intron XP_006712191.1
XM_006712129.1 370 Intron XP_006712192.1
XM_006712130.1 370 Intron XP_006712193.1
XM_011533146.2 370 Intron XP_011531448.1
XM_017005216.1 370 Intron XP_016860705.1

View Full Product Details