Product Details

SNP ID
rs201640210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166198690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGCTCTATTTTTTGCTTTCCTT[C/G]CTGTCTTTCCCTTTGTCTTCCTTTT
Phenotype
MIM: 603415
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 6224 Missense Mutation AGC,AGG S1972R NP_002968.1
XM_005246757.2 6224 Missense Mutation AGC,AGG S1983R XP_005246814.1
XM_011511616.2 6224 Missense Mutation AGC,AGG S1983R XP_011509918.1
XM_011511617.2 6224 Missense Mutation AGC,AGG S1983R XP_011509919.1
XM_011511618.2 6224 Missense Mutation AGC,AGG S1972R XP_011509920.1
XM_011511619.2 6224 Intron XP_011509921.1
XM_017004668.1 6224 Missense Mutation AGC,AGG S1854R XP_016860157.1
XM_017004669.1 6224 Missense Mutation AGC,AGG S1735R XP_016860158.1

View Full Product Details