Product Details

SNP ID
rs201640710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85754830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCGCGTCGCCTAGGAAACGAC[C/T]GGGCGAAGCGACTGCCGCCTCCTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATOH8 PubMed Links

Gene Details

Gene
ATOH8
Gene Name
atonal bHLH transcription factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032827.6 1045 Missense Mutation CCG,CTG P214L NP_116216.2
XM_006712122.3 1045 Missense Mutation CCG,CTG P214L XP_006712185.1
XM_011533139.1 1045 Missense Mutation CCG,CTG P214L XP_011531441.1

View Full Product Details