Product Details

SNP ID
rs79724453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:75732121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAAAACTTGCTTCTTGCTTCCA[A/C]CTTTGGGAAGGAAAAAGGAAAACCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR4273 PubMed Links

Gene Details

Gene
MIR4273
Gene Name
microRNA 4273
There are no transcripts associated with this gene.

Gene
ZNF717
Gene Name
zinc finger protein 717
There are no transcripts associated with this gene.

View Full Product Details