Product Details

SNP ID
rs199499595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48629955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCAGGTTCAGCAAGATGGTGGC[C/T]GTGAAGGTCACCAGCCAGATAAGCT
Phenotype
MIM: 604264 MIM: 610068
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELSR3 PubMed Links

Gene Details

Gene
CELSR3
Gene Name
cadherin EGF LAG seven-pass G-type receptor 3
There are no transcripts associated with this gene.

Gene
MIR6824
Gene Name
microRNA 6824
There are no transcripts associated with this gene.

Gene
SLC26A6
Gene Name
solute carrier family 26 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040454.1 1546 Silent Mutation ACA,ACG T461T NP_001035544.1
NM_001281732.1 1546 Silent Mutation ACA,ACG T446T NP_001268661.1
NM_001281733.1 1546 Silent Mutation ACA,ACG T375T NP_001268662.1
NM_022911.2 1546 Silent Mutation ACA,ACG T482T NP_075062.2
NM_134263.2 1546 Silent Mutation ACA,ACG T482T NP_599025.2
NM_134426.2 1546 Silent Mutation ACA,ACG T482T NP_602298.2
Gene
TMEM89
Gene Name
transmembrane protein 89
There are no transcripts associated with this gene.

View Full Product Details