Product Details

SNP ID
rs199566978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39143353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTGAGATCCACTGAGCTACTC[C/T]GGCCAGCGTCCATGCTGGGTGGCAC
Phenotype
MIM: 606458 MIM: 611430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSRNP1 PubMed Links

Gene Details

Gene
CSRNP1
Gene Name
cysteine and serine rich nuclear protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320559.1 1686 Missense Mutation CAG,CGG Q511R NP_001307488.1
NM_001320560.1 1686 Missense Mutation CAG,CGG Q491R NP_001307489.1
NM_033027.3 1686 Missense Mutation CAG,CGG Q491R NP_149016.2
XM_017007048.1 1686 Missense Mutation CAG,CGG Q491R XP_016862537.1
XM_017007049.1 1686 Missense Mutation CAG,CGG Q231R XP_016862538.1
Gene
MIR6822
Gene Name
microRNA 6822
There are no transcripts associated with this gene.

Gene
TTC21A
Gene Name
tetratricopeptide repeat domain 21A
There are no transcripts associated with this gene.

View Full Product Details