Product Details
- SNP ID
-
rs201872482
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:51863017 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTAGAATAAGAACAAAAGCAGCT[C/G]TAAAGATCCAGGCCTGGTGGCGGGG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IQCF2
PubMed Links
Gene Details
- Gene
- IQCF2
- Gene Name
- IQ motif containing F2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_203424.1 |
180 |
Missense Mutation |
CTA,GTA |
L48V |
NP_982248.1 |
View Full Product Details