Product Details

SNP ID
rs202132773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:195867255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTGTGGTCACCCCATGCACCA[C/T]GGCCTGCAGCTGGGCACACCCACCC
Phenotype
MIM: 606994
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929697 PubMed Links

Gene Details

Gene
LOC101929697
Gene Name
uncharacterized LOC101929697
There are no transcripts associated with this gene.

Gene
TNK2
Gene Name
tyrosine kinase non receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010938.1 3439 Intron NP_001010938.1
NM_001308046.1 3439 Intron NP_001294975.1
NM_005781.4 3439 Missense Mutation NP_005772.3
XM_005269268.3 3439 Missense Mutation XP_005269325.1
XM_005269270.3 3439 Missense Mutation XP_005269327.1
XM_005269274.3 3439 Missense Mutation XP_005269331.1
XM_005269275.3 3439 Missense Mutation XP_005269332.1
XM_011512317.2 3439 Missense Mutation XP_011510619.1
XM_011512318.2 3439 Missense Mutation XP_011510620.2
XM_011512320.1 3439 Missense Mutation XP_011510622.1
XM_011512321.2 3439 Missense Mutation XP_011510623.1
XM_017005508.1 3439 Missense Mutation XP_016860997.1
XM_017005509.1 3439 Missense Mutation XP_016860998.1
XM_017005510.1 3439 Intron XP_016860999.1

View Full Product Details