Product Details

SNP ID
rs202235581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49012110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGGTCTGGTACCCAGCAACTGC[C/G]TTAGCAGACAACAAACCTGTAGCAC
Phenotype
MIM: 615150 MIM: 612911 MIM: 614584 MIM: 606031
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DALRD3 PubMed Links

Gene Details

Gene
DALRD3
Gene Name
DALR anticodon binding domain containing 3
There are no transcripts associated with this gene.

Gene
MIR191
Gene Name
microRNA 191
There are no transcripts associated with this gene.

Gene
MIR425
Gene Name
microRNA 425
There are no transcripts associated with this gene.

Gene
NDUFAF3
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
There are no transcripts associated with this gene.

Gene
P4HTM
Gene Name
prolyl 4-hydroxylase, transmembrane
There are no transcripts associated with this gene.

Gene
WDR6
Gene Name
WD repeat domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320546.1 531 Silent Mutation GCC,GCG A166A NP_001307475.1
NM_001320547.1 531 Silent Mutation GCC,GCG A141A NP_001307476.1
NM_018031.4 531 Silent Mutation GCC,GCG A222A NP_060501.3

View Full Product Details