Product Details

SNP ID
rs200833787
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49108462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTGGGGGCTGGCCGATCCACAG[G/T]GGGGGCGAAGCGTTCAGGGGCTGTC
Phenotype
MIM: 603727 MIM: 614471
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MIR6890 PubMed Links

Gene Details

Gene
MIR6890
Gene Name
microRNA 6890
There are no transcripts associated with this gene.

Gene
QARS
Gene Name
glutaminyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
USP19
Gene Name
ubiquitin specific peptidase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199160.1 4368 Intron NP_001186089.1
NM_001199161.1 4368 Missense Mutation NP_001186090.1
NM_001199162.1 4368 Missense Mutation NP_001186091.1
NM_006677.2 4368 Intron NP_006668.1
XM_005264823.2 4368 Intron XP_005264880.1
XM_005264824.2 4368 Intron XP_005264881.1
XM_005264825.2 4368 Intron XP_005264882.1
XM_005264826.2 4368 Intron XP_005264883.1
XM_005264827.2 4368 Intron XP_005264884.1
XM_005264829.2 4368 Missense Mutation XP_005264886.1
XM_005264830.2 4368 Intron XP_005264887.1
XM_005264831.2 4368 Missense Mutation XP_005264888.1
XM_006712946.2 4368 Intron XP_006713009.1
XM_006712947.2 4368 Intron XP_006713010.1
XM_006712948.2 4368 Missense Mutation XP_006713011.1
XM_006712949.2 4368 Intron XP_006713012.1
XM_006712950.2 4368 Missense Mutation XP_006713013.1
XM_006712951.2 4368 Missense Mutation XP_006713014.1
XM_006712952.2 4368 Missense Mutation XP_006713015.1
XM_006712953.1 4368 Intron XP_006713016.1
XM_017005615.1 4368 Intron XP_016861104.1
XM_017005616.1 4368 Intron XP_016861105.1
XM_017005617.1 4368 Intron XP_016861106.1
XM_017005618.1 4368 Missense Mutation XP_016861107.1
XM_017005619.1 4368 Missense Mutation XP_016861108.1
XM_017005620.1 4368 Missense Mutation XP_016861109.1
XM_017005621.1 4368 Intron XP_016861110.1
XM_017005622.1 4368 Missense Mutation XP_016861111.1
XM_017005623.1 4368 Intron XP_016861112.1
XM_017005624.1 4368 Missense Mutation XP_016861113.1
XM_017005625.1 4368 Intron XP_016861114.1
XM_017005626.1 4368 Intron XP_016861115.1
XM_017005627.1 4368 Intron XP_016861116.1
XM_017005628.1 4368 Intron XP_016861117.1
XM_017005629.1 4368 Intron XP_016861118.1
XM_017005630.1 4368 Missense Mutation XP_016861119.1
XM_017005631.1 4368 Missense Mutation XP_016861120.1
XM_017005632.1 4368 Missense Mutation XP_016861121.1
XM_017005633.1 4368 Missense Mutation XP_016861122.1
XM_017005634.1 4368 Missense Mutation XP_016861123.1
XM_017005635.1 4368 Intron XP_016861124.1

View Full Product Details