Product Details

SNP ID
rs201338202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49832021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGCATTGAGATCAATATCATCA[C/T]GGAAGGATGGCCGGCGGAGCTTCAG
Phenotype
MIM: 605958
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAIP PubMed Links

Gene Details

Gene
TRAIP
Gene Name
TRAF interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005879.2 766 Missense Mutation CAT,CGT H311R NP_005870.2
XM_011533264.1 766 Missense Mutation CAT,CGT H234R XP_011531566.1
XM_017005526.1 766 Missense Mutation CAT,CGT H212R XP_016861015.1

View Full Product Details