Product Details

SNP ID
rs201727979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:158646184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTAGGTGAAAGGTAAAGATGGAG[C/T]TGGTGCTGTCATGGATTCCATGGAA
Phenotype
MIM: 606639
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFM1 PubMed Links

Gene Details

Gene
GFM1
Gene Name
G elongation factor mitochondrial 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308164.1 392 Missense Mutation GCT,GTT A85V NP_001295093.1
NM_001308166.1 392 Missense Mutation GCT,GTT A85V NP_001295095.1
NM_024996.5 392 Missense Mutation GCT,GTT A85V NP_079272.4
XM_006713795.1 392 Missense Mutation GCT,GTT A85V XP_006713858.1

View Full Product Details