Product Details

SNP ID
rs200723262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:20040629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCTGCCGCCGGGGGCTCGGGCG[C/G]CTGCGGTCCGGCGACGGCAGTGGCT
Phenotype
MIM: 602303
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KAT2B PubMed Links

Gene Details

Gene
KAT2B
Gene Name
lysine acetyltransferase 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003884.4 223 Missense Mutation GCC,GGC A51G NP_003875.3
XM_005265528.4 223 Missense Mutation GCC,GGC A51G XP_005265585.1
XM_017007423.1 223 Intron XP_016862912.1
XM_017007424.1 223 Intron XP_016862913.1

View Full Product Details