Product Details

SNP ID
rs200877703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:12568901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTATGCATGGTGTGTGTCGGG[A/C]AGGAAGTCAGTGCCTATTCTCACAT
Phenotype
MIM: 608426
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MKRN2 PubMed Links

Gene Details

Gene
MKRN2
Gene Name
makorin ring finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271707.1 190 Intron NP_001258636.1
NM_014160.4 190 Missense Mutation GAA,GCA E18A NP_054879.3
Gene
MKRN2OS
Gene Name
MKRN2 opposite strand
There are no transcripts associated with this gene.

View Full Product Details