Product Details

SNP ID
rs200692779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44651167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGAAAACTTCCCAGGTCAGGCA[G/T]CCAGCCAACAAGTGCACTCCGAGAA
Phenotype
MIM: 194533
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF197 PubMed Links

Gene Details

Gene
ZNF197
Gene Name
zinc finger protein 197
There are no transcripts associated with this gene.

Gene
ZNF35
Gene Name
zinc finger protein 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003420.3 Intron NP_003411.3
XM_017007129.1 Intron XP_016862618.1
XM_017007130.1 Intron XP_016862619.1
XM_017007131.1 Intron XP_016862620.1

View Full Product Details