Product Details

SNP ID
rs200981578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129252918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTACTTGACCATCAAGGAGATG[A/T]CTTGCATTGCAGAGGATGTCATCAT
Phenotype
MIM: 615525
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COPG1 PubMed Links

Gene Details

Gene
COPG1
Gene Name
coatomer protein complex subunit gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016128.3 386 Missense Mutation ACT,TCT T96S NP_057212.1
XM_011512549.2 386 Missense Mutation ACT,TCT T96S XP_011510851.1

View Full Product Details