Product Details

SNP ID
rs200007633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114128748 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGATCTTGAGGAAGGCTTTCC[A/G]GAACTCGATATTGAAGGTGGTATAG
Phenotype
MIM: 126451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRD3 PubMed Links

Gene Details

Gene
DRD3
Gene Name
dopamine receptor D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000796.5 1594 Missense Mutation CGG,TGG R391W NP_000787.2
NM_001282563.2 1594 Missense Mutation CGG,TGG R391W NP_001269492.1
NM_001290809.1 1594 Missense Mutation CGG,TGG R391W NP_001277738.1
NM_033663.5 1594 Missense Mutation CGG,TGG R358W NP_387512.3
XM_017005829.1 1594 Missense Mutation CGG,TGG R391W XP_016861318.1

View Full Product Details