Product Details

SNP ID
rs200025847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120750598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGAAGTTCCAGCAGCATTGAAG[C/T]GGTTAGCCAAGTATGTGATCCGGGG
Phenotype
MIM: 189962
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF2E1 PubMed Links

Gene Details

Gene
GTF2E1
Gene Name
general transcription factor IIE subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005513.2 231 Missense Mutation CGG,TGG R16W NP_005504.2
XM_011512744.2 231 Missense Mutation CGG,TGG R16W XP_011511046.1
XM_011512745.2 231 Missense Mutation CGG,TGG R16W XP_011511047.1
Gene
RABL3
Gene Name
RAB, member of RAS oncogene family like 3
There are no transcripts associated with this gene.

View Full Product Details