Product Details

SNP ID
rs200183150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50571257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGGTCAGCCACTGGACTGGGGT[A/C]TTTGAGAAGAGGGGTATCCCTGAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C3orf18 PubMed Links

Gene Details

Gene
C3orf18
Gene Name
chromosome 3 open reading frame 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171740.2 750 Intron NP_001165211.1
NM_001171741.2 750 Intron NP_001165212.1
NM_001171743.2 750 Intron NP_001165214.2
NM_016210.4 750 Intron NP_057294.2
XM_011533781.2 750 Intron XP_011532083.1
XM_011533782.2 750 Intron XP_011532084.1
XM_011533783.2 750 Intron XP_011532085.1
XM_011533784.2 750 Intron XP_011532086.1
XM_011533785.2 750 Intron XP_011532087.1
XM_011533788.1 750 Intron XP_011532090.1
XM_011533789.1 750 Intron XP_011532091.1
XM_011533790.1 750 Intron XP_011532092.1
XM_017006545.1 750 Intron XP_016862034.1
XM_017006546.1 750 Intron XP_016862035.1
XM_017006547.1 750 Intron XP_016862036.1
Gene
HEMK1
Gene Name
HemK methyltransferase family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317851.1 750 Silent Mutation GTA,GTC V51V NP_001304780.1
NM_016173.4 750 Silent Mutation GTA,GTC V51V NP_057257.1
XM_005265218.3 750 Silent Mutation GTA,GTC V51V XP_005265275.1
XM_011533806.2 750 Silent Mutation GTA,GTC V51V XP_011532108.1
XM_011533807.1 750 Silent Mutation GTA,GTC V51V XP_011532109.1
XM_011533808.1 750 Silent Mutation GTA,GTC V51V XP_011532110.1
XM_011533809.2 750 Silent Mutation GTA,GTC V51V XP_011532111.1
XM_011533810.2 750 Silent Mutation GTA,GTC V51V XP_011532112.1
XM_011533812.2 750 Silent Mutation GTA,GTC V51V XP_011532114.1
XM_011533813.2 750 UTR 5 XP_011532115.1
XM_011533814.2 750 UTR 5 XP_011532116.1

View Full Product Details