Product Details

SNP ID
rs200028476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52813486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCTCCTGGTAATCCAGCCTGC[A/G]CTCTCTGAAATGGAAAGACAGACAG
Phenotype
MIM: 146650 MIM: 600564
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITIH3 PubMed Links

Gene Details

Gene
ITIH3
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
There are no transcripts associated with this gene.

Gene
ITIH4
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166449.1 2697 Missense Mutation CGC,TGC R880C NP_001159921.1
NM_002218.4 2697 Missense Mutation CGC,TGC R910C NP_002209.2

View Full Product Details