Product Details

SNP ID
rs199602127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150666822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCAACACTGATTCTTGCCTGTT[A/G]GCCCATTGCTAGAAAAGTTATAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERICH6 PubMed Links

Gene Details

Gene
ERICH6
Gene Name
glutamate rich 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308234.1 1462 Nonsense Mutation CAA,TAA Q419* NP_001295163.1
NM_152394.4 1462 Nonsense Mutation CAA,TAA Q565* NP_689607.2
XM_005247120.3 1462 Intron XP_005247177.1

View Full Product Details