Product Details

SNP ID
rs200064431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112563476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTATACATTCCTGCTAGGTGCC[A/G]TATTCATTGCTTTAAGCTCAAGTCG
Phenotype
MIM: 609606
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG3 PubMed Links

Gene Details

Gene
ATG3
Gene Name
autophagy related 3
There are no transcripts associated with this gene.

Gene
SLC35A5
Gene Name
solute carrier family 35 member A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017945.3 356 Missense Mutation ATA,GTA I25V NP_060415.1
XM_011512948.1 356 Missense Mutation ATA,GTA I25V XP_011511250.1

View Full Product Details