Product Details

SNP ID
rs200583939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46743996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGTGACAGGTGTTTGAAGGTG[G/T]ACAAGGGCAACGTCACCCATGATGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRSS43 PubMed Links

Gene Details

Gene
PRSS43
Gene Name
protease, serine 43
There are no transcripts associated with this gene.

Gene
PRSS45
Gene Name
protease, serine 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199183.2 258 Silent Mutation GTA,GTC V86V NP_954652.2
Gene
PRSS46
Gene Name
protease, serine 46
There are no transcripts associated with this gene.

View Full Product Details